Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001355.1 | NP_001001355.1 | 1 | 459 |
Gene Symbol: LOC100909392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758664.1 | XP_008756886.1 | 1 | 114 |
Gene Symbol: LOC100909435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749404.1 | XP_003749452.1 | 1 | 459 |