Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044227.1 | NP_001037692.1 | 2 | 242 | ||
XM_006249941.1 | XP_006250003.1 | 3 | 353 | |||
GenBank mRNA | BC088173.1 | - | 2 | 242 | 7323337 | |
FQ218705.1 | - | 2 | 260 | |||
FQ219461.1 | - | 2 | 263 |