Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105963.2 | NP_001099433.1 | 3 | 274 | ||
XM_006250098.2 | XP_006250160.1 | 3 | 391 | |||
GenBank mRNA | BC166895.1 | - | 3 | 274 | 6920474 | |
FQ214536.1 | - | 3 | 258 |