Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011909.1 | NP_001011909.1 | 2 | 483 | ||
XM_006250435.2 | XP_006250497.1 | 2 | 482 | |||
XM_006250439.2 | XP_006250501.1 | 2 | 531 | |||
XM_008769838.1 | XP_008768060.1 | 2 | 482 | |||
XM_008769839.1 | XP_008768061.1 | 2 | 3132 | |||
XM_008769840.1 | XP_008768062.1 | 2 | 567 | |||
XM_008769841.1 | XP_008768063.1 | 3 | 673 | |||
XM_008769842.1 | XP_008768064.1 | 2 | 482 | |||
XM_008769843.1 | XP_008768065.1 | 2 | 482 | |||
XM_008769844.1 | XP_008768066.1 | 2 | 482 | |||
XM_008769845.1 | XP_008768067.1 | 2 | 482 | |||
XM_008769846.1 | XP_008768068.1 | 2 | 670 | |||
GenBank mRNA | BC079232.1 | - | 2 | 483 | 7132530 | |
FQ230498.1 | - | 2 | 531 |