Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105994.1 | NP_001099464.1 | 2, 3 | 265 | ||
XM_006250541.1 | XP_006250603.1 | 2, 3 | 318 | |||
XM_008769927.1 | XP_008768149.1 | 2, 3 | 1177 | |||
GenBank mRNA | FQ212126.1 | - | 2, 3 | 289 | ||
FQ213206.1 | - | 2, 3 | 191 |