Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006961.1 | NP_001006962.1 | 3 | 510 | ||
XM_006251199.1 | XP_006251261.1 | 2 | 2073 | |||
XM_006251200.2 | XP_006251262.1 | 3 | 1426 | |||
GenBank mRNA | AB167521.1 | - | 3 | 510 |