Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100507.1 | NP_001093977.1 | 3364 | |||
XM_006251439.2 | XP_006251501.1 | 6798 | ||||
XM_006251440.2 | XP_006251502.1 | 6780 | ||||
XM_008770262.1 | XP_008768484.1 | 7600 | ||||
XM_008770263.1 | XP_008768485.1 | 3526 | ||||
XM_008770264.1 | XP_008768486.1 | 7522 | ||||
XM_008770265.1 | XP_008768487.1 | 7504 | ||||
XM_008770266.1 | XP_008768488.1 | 3529 | ||||
XM_008770267.1 | XP_008768489.1 | 3658 | ||||
XM_008770268.1 | XP_008768490.1 | 3462 | ||||
XM_008770269.1 | XP_008768491.1 | 6702 | ||||
GenBank mRNA | BC079401.1 | - | 1214 | |||
BC099192.1 | - | 3 | 384 |
Gene Symbol: LOC103693776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770394.1 | XP_008768616.1 | 18 | 7600 | ||
XM_008770395.1 | XP_008768617.1 | 17 | 3526 | |||
XM_008770396.1 | XP_008768618.1 | 19 | 6876 | |||
XM_008770397.1 | XP_008768619.1 | 17 | 7522 | |||
XM_008770398.1 | XP_008768620.1 | 18 | 7504 | |||
XM_008770399.1 | XP_008768621.1 | 18 | 6798 | |||
XM_008770400.1 | XP_008768622.1 | 19 | 6780 | |||
XM_008770401.1 | XP_008768623.1 | 16 | 3529 | |||
XM_008770402.1 | XP_008768624.1 | 16 | 3658 | |||
XM_008770403.1 | XP_008768625.1 | 17 | 3462 | |||
XM_008770404.1 | XP_008768626.1 | 18 | 6702 |