Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024866.1 | NP_001020037.1 | 2 | 272 | ||
XM_006251598.2 | XP_006251660.1 | 2 | 366 | |||
GenBank mRNA | BC097425.1 | - | 2 | 272 | 7455112 | |
FQ213878.1 | - | 2 | 109 |