Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106027.1 | NP_001099497.1 | 8 | 720 | ||
XM_006251752.2 | XP_006251814.1 | 8 | 1425 | |||
XM_006251753.2 | XP_006251815.1 | 8 | 745 | |||
XM_008770502.1 | XP_008768724.1 | 3 | 1678 | |||
XM_008770503.1 | XP_008768725.1 | 6 | 1131 | |||
XM_008770504.1 | XP_008768726.1 | 7 | 849 | |||
XM_008770505.1 | XP_008768727.1 | 8 | 2353 |