Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017376.1 | NP_001017376.1 | 1261 | |||
XM_006252293.2 | XP_006252355.1 | 5 | 1767 | |||
GenBank mRNA | BC092627.1 | - | 1261 | 7314255 |
Gene Symbol: NEWGENE_1305790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252260.2 | XP_006252322.1 | 5 | 1767 | ||
XM_008770837.1 | XP_008769059.1 | 5 | 1806 |