Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106048.1 | NP_001099518.1 | 3 | 525 | ||
XM_006252262.1 | XP_006252324.1 | 4 | 1294 | |||
XM_006252264.2 | XP_006252326.1 | 4 | 1755 | |||
XM_008770826.1 | XP_008769048.1 | 3 | 2729 |