Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025634.1 | NP_001020805.1 | 4 | 542 | ||
XM_006252512.2 | XP_006252574.1 | 4 | 1875 | |||
XM_006252513.2 | XP_006252575.1 | 4 | 745 | |||
XM_006252514.2 | XP_006252576.1 | 4 | 937 | |||
XM_006252515.2 | XP_006252577.1 | 4 | 995 | |||
XM_006252516.2 | XP_006252578.1 | 3 | 1791 | |||
XM_006252520.2 | XP_006252582.1 | 4 | 848 | |||
XM_006252521.2 | XP_006252583.1 | 3 | 661 | |||
XM_006252522.2 | XP_006252584.1 | 3 | 853 | |||
XM_008770970.1 | XP_008769192.1 | 4 | 1881 | |||
XM_008770971.1 | XP_008769193.1 | 3 | 2186 | |||
XM_008770972.1 | XP_008769194.1 | 3 | 1285 | |||
GenBank mRNA | BC098699.1 | - | 4 | 542 | 7386922 |