Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106106.1 | NP_001099576.1 | 3, 4 | 405 | ||
GenBank mRNA | BC166841.1 | - | 393 | 9027959 | ||
FQ212823.1 | - | 3, 4 | 392 | |||
FQ219292.1 | - | 3, 4 | 424 | |||
FQ220783.1 | - | 3, 4 | 406 | |||
FQ223579.1 | - | 3, 4 | 392 | |||
FQ225943.1 | - | 3, 4 | 424 | |||
FQ229476.1 | - | 3, 4 | 424 | |||
FQ231336.1 | - | 3, 4 | 392 | |||
FQ233549.1 | - | 3, 4 | 392 | |||
FQ234239.1 | - | 3, 4 | 404 |