Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106125.1 | NP_001099595.1 | 3 | 138 | ||
XM_006254307.2 | XP_006254369.1 | 4 | 4052 |