Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010920.1 | NP_001010920.1 | 9 | 1104 | ||
XM_006254998.2 | XP_006255060.1 | 8 | 1150 | |||
XM_006254999.2 | XP_006255061.1 | 9 | 1144 | |||
GenBank mRNA | AY569013.1 | - | 9 | 1104 | ||
BC095904.1 | - | 8 | 1077 | 7110846 |