Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772216.1 | XP_008770438.1 | 7, 8 | 3322 |
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170469.1 | NP_001163940.1 | 9, 10 | 1179 |
Gene Symbol: LOC103690064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772178.1 | XP_008770400.1 | 7, 8 | 1319 |