Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106165.1 | NP_001099635.1 | 7 | 858 | ||
XM_006255051.2 | XP_006255113.1 | 7 | 986 | |||
XM_008772243.1 | XP_008770465.1 | 6 | 724 | |||
GenBank mRNA | BC167038.1 | - | 7 | 827 | 9090629 |