Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106174.1 | NP_001099644.1 | 3 | 649 | ||
XM_006255398.2 | XP_006255460.1 | 2 | 331 | |||
XM_006255399.2 | XP_006255461.1 | 2 | 2470 |