Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106188.1 | NP_001099658.1 | 6 | 665 | ||
XM_006255666.2 | XP_006255728.1 | 6 | 765 | |||
XM_006255670.2 | XP_006255732.1 | 6 | 761 | |||
XM_006255671.2 | XP_006255733.1 | 6 | 761 | |||
XM_006255673.2 | XP_006255735.1 | 6 | 761 | |||
XM_008772491.1 | XP_008770713.1 | 6 | 766 |