Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019166.2 | NP_062039.1 | 4 | 594 | ||
GenBank mRNA | U39549.1 | - | 4 | 563 |