Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019170.2 | NP_062043.1 | 575 | |||
GenBank mRNA | D89069.1 | - | 568 | |||
X84349.1 | - | 575 |
Gene Symbol: LOC100360507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751016.3 | XP_003751064.2 | 3 | 685 |
Gene Symbol: LOC102556347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248109.2 | XP_006248171.1 | 685 |