Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017138.2 | NP_058834.1 | 4 | 360 | ||
XM_008766672.1 | XP_008764894.1 | 4 | 433 | |||
GenBank mRNA | BC060578.1 | - | 4 | 378 | 6919235 | |
D25224.1 | - | 4 | 361 | |||
FJ386454.1 | - | 3 | 341 | |||
FQ209463.1 | - | 4 | 374 | |||
FQ209469.1 | - | 4 | 374 | |||
FQ209490.1 | - | 4 | 375 | |||
FQ209667.1 | - | 4 | 374 | |||
FQ209729.1 | - | 4 | 374 | |||
FQ211189.1 | - | 4 | 374 | |||
FQ212660.1 | - | 4 | 374 | |||
FQ213275.1 | - | 4 | 374 | |||
FQ213317.1 | - | 4 | 374 | |||
FQ213334.1 | - | 4 | 374 | |||
FQ213623.1 | - | 4 | 377 | |||
FQ213835.1 | - | 4 | 375 | |||
FQ213894.1 | - | 4 | 374 | |||
FQ215242.1 | - | 4 | 374 | |||
FQ215292.1 | - | 4 | 375 | |||
FQ216063.1 | - | 4 | 374 | |||
FQ216085.1 | - | 4 | 374 | |||
FQ216130.1 | - | 4 | 374 | |||
FQ216245.1 | - | 4 | 374 | |||
FQ216788.1 | - | 4 | 376 | |||
FQ216997.1 | - | 4 | 374 | |||
FQ217664.1 | - | 4 | 374 | |||
FQ219586.1 | - | 4 | 374 | |||
FQ219785.1 | - | 4 | 377 | |||
FQ220012.1 | - | 4 | 374 | |||
FQ220017.1 | - | 4 | 372 | |||
FQ220075.1 | - | 4 | 375 | |||
FQ220084.1 | - | 4 | 377 | |||
FQ220126.1 | - | 4 | 374 | |||
FQ220134.1 | - | 4 | 374 | |||
FQ220171.1 | - | 4 | 373 | |||
FQ220184.1 | - | 4 | 374 | |||
FQ220347.1 | - | 4 | 376 | |||
FQ220461.1 | - | 4 | 355 | |||
FQ220468.1 | - | 4 | 374 | |||
FQ220661.1 | - | 4 | 377 | |||
FQ220822.1 | - | 4 | 374 | |||
FQ220904.1 | - | 4 | 375 | |||
FQ220964.1 | - | 4 | 374 | |||
FQ221103.1 | - | 4 | 374 | |||
FQ221334.1 | - | 4 | 394 | |||
FQ221443.1 | - | 4 | 375 | |||
FQ221672.1 | - | 4 | 374 | |||
FQ222093.1 | - | 4 | 376 | |||
FQ222280.1 | - | 4 | 374 | |||
FQ222542.1 | - | 4 | 376 | |||
FQ222757.1 | - | 4 | 372 | |||
FQ223223.1 | - | 4 | 374 | |||
FQ224747.1 | - | 4 | 374 | |||
FQ225510.1 | - | 4 | 375 | |||
FQ226054.1 | - | 4 | 374 | |||
FQ226408.1 | - | 4 | 374 | |||
FQ227293.1 | - | 4 | 374 | |||
FQ227600.1 | - | 4 | 375 | |||
FQ229290.1 | - | 4 | 376 | |||
FQ229305.1 | - | 4 | 376 | |||
FQ229367.1 | - | 4 | 375 | |||
FQ229516.1 | - | 4 | 374 | |||
FQ229579.1 | - | 4 | 374 | |||
FQ229598.1 | - | 4 | 375 | |||
FQ229712.1 | - | 4 | 374 | |||
FQ229787.1 | - | 4 | 374 | |||
FQ229850.1 | - | 4 | 376 | |||
FQ229859.1 | - | 4 | 372 | |||
FQ229918.1 | - | 4 | 394 | |||
FQ229945.1 | - | 4 | 374 | |||
FQ229970.1 | - | 4 | 374 | |||
FQ230088.1 | - | 4 | 374 | |||
FQ230106.1 | - | 4 | 374 | |||
FQ230180.1 | - | 4 | 375 | |||
FQ230258.1 | - | 4 | 373 | |||
FQ231469.1 | - | 3 | 191 | |||
FQ231622.1 | - | 4 | 372 | |||
FQ232305.1 | - | 4 | 376 | |||
FQ232568.1 | - | 4 | 375 | |||
FQ233068.1 | - | 4 | 374 | |||
FQ233112.1 | - | 4 | 374 | |||
FQ233340.1 | - | 4 | 376 | |||
FQ233547.1 | - | 4 | 375 | |||
FQ233919.1 | - | 4 | 374 | |||
FQ234511.1 | - | 4 | 374 | |||
FQ234742.1 | - | 4 | 374 | |||
FQ234765.1 | - | 4 | 374 | |||
FQ234772.1 | - | 4 | 374 |