Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169143.1 | NP_001162614.1 | 2 | 101 | ||
XM_006228186.2 | XP_006228248.1 | 3 | 646 | |||
XM_008758902.1 | XP_008757124.1 | 3 | 666 | |||
XM_008758903.1 | XP_008757125.1 | 2 | 151 | |||
GenBank mRNA | FM030255.1 | - | 2 | 90 |