Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106226.2 | NP_001099696.2 | 7, 8 | 3102 | ||
XM_006228343.2 | XP_006228405.1 | 9, 10 | 3449 | |||
XM_006228344.2 | XP_006228406.1 | 8, 9 | 3383 | |||
XM_006228345.2 | XP_006228407.1 | 8, 9 | 3272 |