Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009283.1 | NP_001009283.1 | 4, 5 | 780 | ||
XM_006228866.2 | XP_006228928.1 | 5, 6 | 1228 | |||
XM_006228867.1 | XP_006228929.1 | 5, 6 | 1015 | |||
XM_006228868.2 | XP_006228930.1 | 4, 5 | 1084 | |||
GenBank mRNA | BC088237.1 | - | 4, 5 | 780 | 7325652 |
Gene Symbol: LOC100911963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748821.2 | XP_003748869.1 | 4, 5 | 871 | ||
XM_003748822.2 | XP_003748870.1 | 5, 6 | 1015 | |||
XM_006228801.2 | XP_006228863.1 | 5, 6 | 1229 | |||
XM_006228802.2 | XP_006228864.1 | 4, 5 | 1085 |