Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032613.2 | NP_116002.1 | 1 | 140 | ||
GenBank mRNA | AF242187.1 | - | 1 | 54 | ||
BC099791.1 | - | 1 | 140 | 7934892 |