Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005382.1 | NP_001005382.1 | 5 | 693 |
Gene Symbol: LOC687880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229168.1 | XP_006229230.1 | 699 |