Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1309036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106255.1 | NP_001099725.1 | 5 | 979 | ||
XM_006228989.2 | XP_006229051.1 | 5 | 1161 | |||
XM_008759332.1 | XP_008757554.1 | 4 | 778 |