Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170426.1 | NP_001163897.1 | 16 | 1992 | ||
XM_006229007.2 | XP_006229069.1 | 16 | 2245 | |||
XM_006229008.2 | XP_006229070.1 | 16 | 2242 | |||
XM_006229009.2 | XP_006229071.1 | 15 | 2174 | |||
XM_006229010.2 | XP_006229072.1 | 15 | 2140 | |||
XM_006229011.2 | XP_006229073.1 | 16 | 2240 | |||
XM_006229012.2 | XP_006229074.1 | 15 | 2172 | |||
XM_006229013.2 | XP_006229075.1 | 15 | 1854 |