Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034127.1 | NP_001029299.1 | 9 | 1026 | ||
GenBank mRNA | BC100614.1 | - | 9 | 1026 | 5598102 |