Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106284.1 | NP_001099754.1 | 3, 4 | 327 | ||
XM_006229870.2 | XP_006229932.1 | 2, 3 | 551 | |||
XM_006229871.2 | XP_006229933.1 | 2, 3 | 438 | |||
XM_006229872.2 | XP_006229934.1 | 2, 3 | 185 | |||
XM_006229873.2 | XP_006229935.1 | 2, 3 | 227 | |||
XM_006229874.2 | XP_006229936.1 | 3, 4 | 283 | |||
XM_006229875.2 | XP_006229937.1 | 2, 3 | 153 | |||
GenBank mRNA | EU627092.1 | - | 2, 3 | 196 |