Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024131.1 | NP_077045.1 | 2 | 219 | ||
GenBank mRNA | FQ209890.1 | - | 2 | 236 | ||
FQ209898.1 | - | 2 | 191 | |||
FQ210467.1 | - | 2 | 192 | |||
FQ210521.1 | - | 2 | 235 | |||
FQ220991.1 | - | 2 | 229 | |||
Z36980.1 | - | 2 | 219 |