Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024131.1 | NP_077045.1 | 2, 3 | 364 | ||
GenBank mRNA | FQ209890.1 | - | 2, 3 | 381 | ||
FQ209898.1 | - | 2, 3 | 336 | |||
FQ210467.1 | - | 2, 3 | 337 | |||
FQ210521.1 | - | 2, 3 | 380 | |||
FQ220991.1 | - | 2, 3 | 374 | |||
Z36980.1 | - | 2, 3 | 364 |