Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024131.1 | NP_077045.1 | 1 | 88 | ||
GenBank mRNA | FQ209890.1 | - | 1 | 105 | ||
FQ209898.1 | - | 1 | 60 | |||
FQ210467.1 | - | 1 | 61 | |||
FQ210521.1 | - | 1 | 104 | |||
FQ220991.1 | - | 1 | 98 | |||
Z36980.1 | - | 1 | 88 |