Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017167.2 | NP_058863.1 | 4 | 817 | ||
XM_008763523.1 | XP_008761745.1 | 3 | 947 | |||
GenBank mRNA | D16534.1 | - | 3 | 795 | ||
D16829.1 | - | 3 | 857 | |||
L22001.1 | - | 4 | 817 | |||
L22536.1 | - | 4 | 969 | |||
U00442.1 | - | 4 | 1003 |
Set Membership: |
G-protein coupled receptor |