Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017173.1 | NP_058869.1 | 2 | 135 | ||
GenBank mRNA | BC086529.1 | - | 1 | 26 | 7303866 | |
M69246.1 | - | 2 | 135 |