Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106299.1 | NP_001099769.1 | 3 | 269 | ||
GenBank mRNA | BC157822.1 | - | 3 | 193 | 8374205 | |
FQ225822.1 | - | 3 | 190 |