Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198737.2 | NP_942032.1 | 2, 3 | 224 | ||
GenBank mRNA | AY316590.1 | - | 2, 3 | 225 | ||
BC099071.1 | - | 2, 3 | 224 | 7381266 |