Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106320.2 | NP_001099790.2 | 25, 26 | 3537 | ||
XM_006230718.2 | XP_006230780.1 | 29, 30 | 3733 | |||
XM_006230723.2 | XP_006230785.1 | 25, 26 | 3538 |
Gene Symbol: LOC103689951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230627.2 | XP_006230689.1 | 8, 9 | 1040 |