Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019195.2 | NP_062068.1 | 5 | 769 | ||
XM_006248262.2 | XP_006248324.1 | 6 | 1462 | |||
XM_006248263.2 | XP_006248325.1 | 6 | 1462 | |||
XM_006248264.2 | XP_006248326.1 | 6 | 1462 | |||
XM_006248265.2 | XP_006248327.1 | 6 | 1462 | |||
XM_006248266.2 | XP_006248328.1 | 5 | 791 | |||
XM_006248267.2 | XP_006248329.1 | 5 | 1348 | |||
XM_006248268.2 | XP_006248330.1 | 6 | 1660 | |||
GenBank mRNA | AF017437.1 | - | 5 | 655 | ||
BC085740.1 | - | 5 | 769 | 7189894 | ||
D87659.1 | - | 5 | 679 | |||
FQ209533.1 | - | 4 | 392 | |||
FQ232147.1 | - | 1 | 23 | |||
FQ232318.1 | - | 4 | 392 |