Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011937.1 | NP_001011937.1 | 9, 10 | 1068 | ||
XM_006230735.2 | XP_006230797.1 | 9, 10 | 2092 | |||
GenBank mRNA | BC083779.1 | - | 9, 10 | 1068 | 7193879 |