Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011943.1 | NP_001011943.1 | 3 | 380 | ||
XM_006231472.1 | XP_006231534.1 | 3 | 491 | |||
XM_006231474.2 | XP_006231536.1 | 3 | 492 | |||
GenBank mRNA | BC079096.1 | - | 3 | 380 | 7124109 |