Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC294154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039607.1 | NP_001034696.1 | 3, 4 | 539 | ||
XM_008772799.1 | XP_008771021.1 | 3, 4 | 733 | |||
XM_008772800.1 | XP_008771022.1 | 3, 4 | 847 | |||
XM_008772801.1 | XP_008771023.1 | 3, 4 | 1272 | |||
GenBank mRNA | BC112320.1 | - | 3, 4 | 539 | 7933524 |