Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133287.1 | NP_579821.1 | 6 | 757 | ||
GenBank mRNA | AB210108.1 | - | 6 | 757 | ||
BC166596.1 | - | 1056 | 7934724 | |||
DQ336702.1 | - | 7 | 791 | |||
DQ336703.1 | - | 6 | 758 | |||
DQ336704.1 | - | 6 | 758 | |||
DQ336705.1 | - | 7 | 791 |