Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-Db1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008884.2 | NP_001008884.1 | 2 | 179 | ||
GenBank mRNA | AF084933.1 | - | 2 | 119 | ||
AJ003231.1 | - | 2 | 64 | |||
AJ003234.1 | - | 2 | 64 | |||
AY626207.1 | - | 2 | 115 | |||
BC087691.1 | - | 2 | 156 | 7310949 | ||
CO563535.1 | - | 2 | 179 | |||
FQ226724.1 | - | 2 | 167 | |||
FQ226759.1 | - | 2 | 173 | |||
FQ227087.1 | - | 2 | 162 | |||
FQ228124.1 | - | 2 | 144 | |||
FQ228129.1 | - | 2 | 167 | |||
FQ228185.1 | - | 2 | 185 | |||
FQ230360.1 | - | 2 | 167 | |||
FQ231387.1 | - | 2 | 167 | |||
FQ231709.1 | - | 2 | 167 | |||
FQ232046.1 | - | 2 | 167 | |||
FQ232100.1 | - | 2 | 167 | |||
FQ232453.1 | - | 2 | 167 | |||
FQ233049.1 | - | 2 | 163 | |||
FQ233335.1 | - | 2 | 167 | |||
FQ233396.1 | - | 2 | 167 | |||
FQ233493.1 | - | 2 | 167 | |||
FQ233800.1 | - | 2 | 145 | |||
FQ233801.1 | - | 2 | 167 | |||
FQ234464.1 | - | 2 | 166 | |||
FQ234907.1 | - | 2 | 167 | |||
KC222950.1 | - | 2 | 115 |