Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164706.1 | NP_001158178.1 | 4, 5 | 382 | ||
NM_001164707.1 | NP_001158179.1 | 3, 4 | 435 | |||
NM_212494.2 | NP_997659.2 | 4, 5 | 435 | |||
XM_006256120.2 | XP_006256182.1 | 5, 6 | 466 | |||
GenBank mRNA | BP501235.1 | - | 3, 4 | 449 | ||
CB586341.1 | - | 4, 5 | 382 | |||
DV717217.1 | - | 3, 4 | 407 | |||
FQ220708.1 | - | 4, 5 | 669 | |||
FQ223106.1 | - | 2, 3 | 109 | |||
FQ229047.1 | - | 2, 3 | 109 |