Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106391.2 | NP_001099861.1 | 2, 3 | 183 | ||
GenBank mRNA | BC158719.1 | - | 2, 3 | 183 | 9089752 |