Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 10 | 1213 | ||
XM_008768268.1 | XP_008766490.1 | 14 | 2386 | |||
XM_008768269.1 | XP_008766491.1 | 13 | 2299 | |||
XM_008768270.1 | XP_008766492.1 | 13 | 2294 | |||
XM_008768271.1 | XP_008766493.1 | 12 | 2212 | |||
XM_008768272.1 | XP_008766494.1 | 13 | 2187 | |||
XM_008768273.1 | XP_008766495.1 | 12 | 2094 | |||
XM_008768274.1 | XP_008766496.1 | 13 | 1620 | |||
XM_008768275.1 | XP_008766497.1 | 12 | 1533 | |||
XM_008768277.1 | XP_008766499.1 | 12 | 1419 | |||
XM_008768278.1 | XP_008766500.1 | 11 | 1332 | |||
XM_008768279.1 | XP_008766501.1 | 11 | 1327 | |||
XM_008768280.1 | XP_008766502.1 | 10 | 1239 | |||
XM_008768281.1 | XP_008766503.1 | 9 | 1152 | |||
XM_008768282.1 | XP_008766504.1 | 15 | 2441 | |||
GenBank mRNA | BC126095.1 | - | 9 | 1120 | 8367931 | |
CO405815.1 | - | 1 | 123 | |||
M84156.1 | - | 13 | 2044 | |||
X79321.1 | - | 10 | 1300 |