Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232783.2 | XP_006232845.1 | 15, 16 | 2733 | ||
XM_006232784.2 | XP_006232846.1 | 16, 17 | 3078 | |||
XM_006232785.2 | XP_006232847.1 | 13, 14 | 2525 | |||
XM_215597.8 | XP_215597.6 | 15, 16 | 2731 |