Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106451.1 | NP_001099921.1 | 1, 2 | 219 | ||
XM_006233027.2 | XP_006233089.1 | 2, 3 | 794 | |||
XM_008761285.1 | XP_008759507.1 | 2, 3 | 5093 |